siRNA Details
| Gene Symbol: Slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135151.1 | NP_001128623.1 | 7 | 1039 | ||
| NM_001135152.1 | NP_001128624.1 | 7 | 1005 | |||
| NM_144808.4 | NP_659057.2 | 7 | 1039 | |||
| GenBank mRNA | AB177995.1 | - | 7 | 1042 | ||
| AK090193.1 | - | 7 | 1002 | |||
| AK192726.1 | - | 2 | 106 | |||
| BC021530.1 | - | 7 | 1042 | 5353717 | ||
| CB521415.1 | - | 4 | 367 | |||

Slc39a14
Genomic Map