siRNA Details
| Gene Symbol: Slc39a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135151.1 | NP_001128623.1 | 3 | 404 | ||
| NM_001135152.1 | NP_001128624.1 | 3 | 370 | |||
| NM_144808.4 | NP_659057.2 | 3 | 404 | |||
| GenBank mRNA | AB177995.1 | - | 3 | 407 | ||
| AK050076.1 | - | 3 | 371 | |||
| AK090193.1 | - | 3 | 369 | |||
| AK122197.1 | - | 1 | 21 | |||
| AK158096.1 | - | 2 | 296 | |||
| BC021530.1 | - | 3 | 407 | 5353717 | ||

Slc39a14
Genomic Map