siRNA Details
| Gene Symbol: Gabpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029885.1 | NP_084161.1 | 6 | 827 | ||
| NM_172512.2 | NP_766100.1 | 5 | 730 | |||
| GenBank mRNA | AK030849.1 | - | 5 | 499 | ||
| AK036692.1 | - | 5 | 1311 | |||
| AK086817.1 | - | 4 | 1391 | |||
| AK087979.1 | - | 6 | 644 | |||
| AK141382.1 | - | 6 | 801 | |||
| AK163939.1 | - | 5 | 1307 | |||
| BC150160.1 | - | 6 | 578 | 40141577 | ||
| BC150482.1 | - | 6 | 578 | 40141578 | ||

Gabpb2
Genomic Map