siRNA Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172513.3 | NP_766101.3 | 12 | 1680 | ||
| GenBank mRNA | AK052638.1 | - | 12 | 1607 | ||
| AK144602.1 | - | 13 | 1785 | |||
| AK168372.1 | - | 12 | 1670 | |||
| BC049806.1 | - | 12 | 1616 | |||
| BC068302.1 | - | 12 | 1588 | 30431671 | ||

Fam126b
Genomic Map