siRNA Details
| Gene Symbol: Fam126b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172513.3 | NP_766101.3 | 9 | 930 | ||
| GenBank mRNA | AK052638.1 | - | 9 | 857 | ||
| AK144602.1 | - | 9 | 867 | |||
| AK168372.1 | - | 9 | 920 | |||
| BC049806.1 | - | 9 | 867 | |||
| BC068302.1 | - | 9 | 838 | 30431671 | ||

Fam126b
Genomic Map