siRNA Details
| Gene Symbol: Wdr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153391.2 | NP_700440.2 | 27 | 3094 | ||
| GenBank mRNA | AK129413.1 | - | 19 | 2183 | ||
| AK147970.1 | - | 27 | 3094 | |||
| AY029258.1 | - | 27 | 3164 | |||
| BC046432.1 | - | 12 | 1296 | |||
| BC060134.1 | - | 27 | 3088 | |||
| BC082301.1 | - | 26 | 2974 | |||

Wdr19
Genomic Map