siRNA Details
| Gene Symbol: Wdr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153391.2 | NP_700440.2 | 31 | 3430 | ||
| GenBank mRNA | AK129413.1 | - | 22 | 3802 | ||
| AK147970.1 | - | 31 | 3430 | |||
| AY029258.1 | - | 31 | 3500 | |||
| BC046432.1 | - | 16 | 1632 | |||
| BC082301.1 | - | 29 | 4862 | |||

Wdr19
Genomic Map