siRNA Details
| Gene Symbol: Wdr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153391.2 | NP_700440.2 | 9 | 877 | ||
| GenBank mRNA | AK043836.1 | - | 9 | 834 | ||
| AK052303.1 | - | 9 | 857 | |||
| AK147970.1 | - | 9 | 877 | |||
| AK166172.1 | - | 9 | 843 | |||
| AY029258.1 | - | 9 | 947 | |||
| BC060134.1 | - | 9 | 871 | |||
| BC082301.1 | - | 8 | 757 | |||

Wdr19
Genomic Map