siRNA Details
| Gene Symbol: Cdkl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166653.1 | NP_001160125.1 | 12 | 1920 | ||
| NM_001166654.1 | NP_001160126.1 | 12 | 2010 | |||
| NM_001166656.1 | NP_001160128.1 | 12 | 1920 | |||
| NM_153785.4 | NP_722480.3 | 12 | 2010 | |||
| GenBank mRNA | AK043995.1 | - | 4 | 604 | ||
| AK045036.1 | - | 11 | 2368 | |||
| AK045356.1 | - | 12 | 1910 | |||
| AK046394.1 | - | 12 | 2011 | |||
| AK078804.1 | - | 12 | 1902 | |||
| AK166428.1 | - | 12 | 1855 | |||
| BB759970.1 | - | 1 | 65 | |||

Cdkl3
Genomic Map