siRNA Details
| Gene Symbol: Zbbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172515.2 | NP_766103.2 | 9, 10 | 1059 | ||
| GenBank mRNA | AK029899.1 | - | 9, 10 | 1052 | ||
| AK133102.1 | - | 9, 10 | 1110 | |||
| BC050789.1 | - | 9, 10 | 1059 | 6706914 | ||
| BC100435.1 | - | 9, 10 | 1059 | |||

Zbbx
Genomic Map