siRNA Details
| Gene Symbol: Zbbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172515.2 | NP_766103.2 | 15 | 1929 | ||
| GenBank mRNA | AK029899.1 | - | 15 | 1922 | ||
| AK133102.1 | - | 15 | 1980 | |||
| BC050789.1 | - | 15 | 1929 | 6706914 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbbx| Gene Symbol: Zbbx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172515.2 | NP_766103.2 | 15 | 1929 | ||
| GenBank mRNA | AK029899.1 | - | 15 | 1922 | ||
| AK133102.1 | - | 15 | 1980 | |||
| BC050789.1 | - | 15 | 1929 | 6706914 | ||