siRNA Details
| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198021.2 | NP_932138.1 | 7 | 1153 | ||
| GenBank mRNA | AK139761.1 | - | 7 | 1139 | ||
| BC042443.1 | - | 7 | 1153 | 4037878 | ||
| BC042770.1 | - | 2 | 52 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scyl2| Gene Symbol: Scyl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198021.2 | NP_932138.1 | 7 | 1153 | ||
| GenBank mRNA | AK139761.1 | - | 7 | 1139 | ||
| BC042443.1 | - | 7 | 1153 | 4037878 | ||
| BC042770.1 | - | 2 | 52 | |||