siRNA Details
| Gene Symbol: Pddc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172116.4 | NP_742114.3 | 3 | 293 | ||
| GenBank mRNA | AK081187.1 | - | 3 | 253 | ||
| AK084273.1 | - | 3 | 249 | |||
| AK088159.1 | - | 3 | 269 | |||
| AK140975.1 | - | 3 | 244 | |||
| AK161205.1 | - | 3 | 259 | |||
| BC023835.1 | - | 3 | 263 | 5345186 | ||

Pddc1
Genomic Map