siRNA Details
| Gene Symbol: Dstyk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172516.4 | NP_766104.2 | 7, 8 | 2102 | ||
| GenBank mRNA | AK043199.1 | - | 2, 3 | 971 | ||
| AK050542.1 | - | 3, 4 | 348 | |||
| AK083696.1 | - | 7, 8 | 2000 | |||
| AK132119.1 | - | 7, 8 | 1977 | |||
| AK145449.1 | - | 7, 8 | 1947 | |||
| AK172954.1 | - | 7, 8 | 1948 | |||
| AY208852.1 | - | 7, 8 | 1949 | |||
| AY429676.1 | - | 7, 8 | 1988 | |||
| BC019133.1 | - | 5, 6 | 737 | |||
| BC089380.1 | - | 4, 5 | 519 | |||

Dstyk
Genomic Map