siRNA Details
| Gene Symbol: Fbxo42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172518.3 | NP_766106.2 | 5 | 814 | ||
| GenBank mRNA | AK028867.1 | - | 5 | 660 | ||
| AK150804.1 | - | 5 | 805 | |||
| AK151503.1 | - | 5 | 771 | |||
| AK165284.1 | - | 5 | 788 | |||
| AK170202.1 | - | 5 | 816 | |||
| AK171215.1 | - | 5 | 817 | |||
| AK208927.1 | - | 3 | 207 | |||
| BC003960.1 | - | 2 | 55 | |||
| BC058667.1 | - | 5 | 805 | 6410012 | ||

Fbxo42
Genomic Map