siRNA Details
| Gene Symbol: Plekhg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198604.2 | NP_941006.2 | 8 | 1180 | ||
| GenBank mRNA | AK153619.1 | - | 8 | 1470 | ||
| BC026778.1 | - | 8 | 1183 | 5133546 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekhg6| Gene Symbol: Plekhg6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198604.2 | NP_941006.2 | 8 | 1180 | ||
| GenBank mRNA | AK153619.1 | - | 8 | 1470 | ||
| BC026778.1 | - | 8 | 1183 | 5133546 | ||