siRNA Details
| Gene Symbol: Plekhh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177606.4 | NP_808274.2 | 8 | 1072 | ||
| GenBank mRNA | AK029252.1 | - | 8 | 1074 | ||
| AK173337.1 | - | 5 | 728 | |||
| BC021518.1 | - | 8 | 1044 | |||
| BC125583.1 | - | 8 | 1037 | 40130098 | ||
| BC137804.1 | - | 8 | 1034 | 8860823 | ||

Plekhh2
Genomic Map