siRNA Details
| Gene Symbol: Dync2li1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172256.1 | NP_758460.1 | 5 | 383 | ||
| GenBank mRNA | AK008822.1 | - | 4 | 332 | ||
| AK160259.1 | - | 5 | 397 | |||
| BC030450.1 | - | 5 | 385 | 5400492 | ||
| BC039070.1 | - | 5 | 383 | 3482503 | ||

Dync2li1
Genomic Map