siRNA Details
| Gene Symbol: Zfp598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183149.1 | NP_898972.1 | 2 | 281 | ||
| GenBank mRNA | AK131127.1 | - | 2 | 300 | ||
| BC024690.1 | - | 2 | 75 | |||
| BC050859.1 | - | 2 | 281 | 6328878 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp598| Gene Symbol: Zfp598 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183149.1 | NP_898972.1 | 2 | 281 | ||
| GenBank mRNA | AK131127.1 | - | 2 | 300 | ||
| BC024690.1 | - | 2 | 75 | |||
| BC050859.1 | - | 2 | 281 | 6328878 | ||