siRNA Details
Gene Symbol: Col28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037865.1 | NP_001032954.1 | 3 | 311 | ||
GenBank mRNA | AJ890449.1 | - | 3 | 311 | ||
BC145537.1 | - | 3 | 298 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Col28a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037865.1 | NP_001032954.1 | 3 | 311 | ||
GenBank mRNA | AJ890449.1 | - | 3 | 311 | ||
BC145537.1 | - | 3 | 298 |