siRNA Details
| Gene Symbol: Tnrc6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144812.2 | NP_659061.2 | 8 | 970 | ||
| NM_177124.4 | NP_796098.3 | 5 | 761 | |||
| GenBank mRNA | AK048628.1 | - | 8 | 970 | ||
| AK051922.1 | - | 5 | 730 | |||
| BC068158.1 | - | 5 | 656 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_138845, XM_006242063, XM_006242061, XM_006242064.

Tnrc6b
Genomic Map