siRNA Details
| Gene Symbol: Dnajc16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172338.2 | NP_758841.1 | 2 | 207 | ||
| GenBank mRNA | AK028692.1 | - | 2 | 212 | ||
| AK051048.1 | - | 2 | 208 | |||
| AK122407.1 | - | 454 | ||||
| AK154946.1 | - | 228 | ||||
| AK164023.1 | - | 2 | 223 | |||

Dnajc16
Genomic Map