siRNA Details
| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172524.3 | NP_766112.1 | 6 | 1136 | ||
| GenBank mRNA | AK035561.1 | - | 6 | 1137 | ||
| AK035736.1 | - | 6 | 1114 | |||
| BC139819.1 | - | 6 | 1127 | 40141293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipal4| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172524.3 | NP_766112.1 | 6 | 1136 | ||
| GenBank mRNA | AK035561.1 | - | 6 | 1137 | ||
| AK035736.1 | - | 6 | 1114 | |||
| BC139819.1 | - | 6 | 1127 | 40141293 | ||