siRNA Details
Gene Symbol: Kmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081049.1 | NP_001074518.1 | 25 | 6248 | ||
GenBank mRNA | AK220457.1 | - | 23 | 3887 | ||
L17069.1 | - | 25 | 5959 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081049.1 | NP_001074518.1 | 25 | 6248 | ||
GenBank mRNA | AK220457.1 | - | 23 | 3887 | ||
L17069.1 | - | 25 | 5959 |