siRNA Details
| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145399.1 | NP_663374.1 | 9, 10 | 809 | ||
| GenBank mRNA | AK148468.1 | - | 9, 10 | 761 | ||
| BC016093.1 | - | 9, 10 | 809 | 4504330 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgn| Gene Symbol: Scgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145399.1 | NP_663374.1 | 9, 10 | 809 | ||
| GenBank mRNA | AK148468.1 | - | 9, 10 | 761 | ||
| BC016093.1 | - | 9, 10 | 809 | 4504330 | ||