siRNA Details
| Gene Symbol: Ttc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172526.3 | NP_766114.1 | 7 | 1339 | ||
| GenBank mRNA | AK039884.1 | - | 7 | 1340 | ||
| AK076743.1 | - | 2 | 155 | |||
| BC055114.1 | - | 2 | 154 | |||
| BC147554.1 | - | 6 | 1043 | 9056545 | ||
| BC147555.1 | - | 6 | 1043 | 9056546 | ||

Ttc24
Genomic Map