siRNA Details
| Gene Symbol: Etnk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175443.5 | NP_780652.2 | 5 | 986 | ||
| GenBank mRNA | AK031665.1 | - | 4 | 2018 | ||
| AK077179.1 | - | 5 | 725 | |||
| AK087818.1 | - | 4 | 784 | |||
| AY212244.1 | - | 5 | 577 | |||
| BC145351.1 | - | 5 | 765 | 9053907 | ||
| BC152310.1 | - | 5 | 924 | 40089976 | ||

Etnk2
Genomic Map