siRNA Details
| Gene Symbol: Zcchc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153538.3 | NP_705766.3 | 4 | 870 | ||
| GenBank mRNA | AK031043.1 | - | 3 | 447 | ||
| BC043111.1 | - | 4 | 824 | 6407010 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zcchc6| Gene Symbol: Zcchc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153538.3 | NP_705766.3 | 4 | 870 | ||
| GenBank mRNA | AK031043.1 | - | 3 | 447 | ||
| BC043111.1 | - | 4 | 824 | 6407010 | ||