siRNA Details
Gene Symbol: Gm4787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 771 | ||
GenBank mRNA | BC141094.1 | - | 1 | 749 | 9055706 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm4787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038995.2 | NP_001034084.2 | 1 | 771 | ||
GenBank mRNA | BC141094.1 | - | 1 | 749 | 9055706 |