siRNA Details
| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146048.1 | NP_001139520.1 | 13 | 1632 | ||
| NM_172528.3 | NP_766116.3 | 13 | 1280 | |||
| GenBank mRNA | AK082721.1 | - | 13 | 1281 | ||
| AK147075.1 | - | 13 | 1632 | |||
| AK220166.1 | - | 13 | 1650 | |||
| BC022613.1 | - | 5 | 574 | |||
| BC046591.1 | - | 13 | 1619 | |||
| BC087542.1 | - | 13 | 1479 | 4934980 | ||

Lrrc1
Genomic Map