siRNA Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177460.4 | NP_803411.3 | 5, 6 | 976 | ||
| GenBank mRNA | AK033791.1 | - | 926 | |||
| AK133475.1 | - | 5, 6 | 977 | |||
| AK166002.1 | - | 4, 5 | 740 | |||
| BC002197.1 | - | 1, 2 | 119 | |||
| BC014731.1 | - | 4, 5 | 1006 | |||
| BC055447.1 | - | 5, 6 | 945 | 3594406 | ||

Parp16
Genomic Map