siRNA Details
| Gene Symbol: Parp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177460.4 | NP_803411.3 | 3 | 567 | ||
| GenBank mRNA | AK033791.1 | - | 517 | |||
| AK133475.1 | - | 3 | 568 | |||
| AK166002.1 | - | 2 | 331 | |||
| AK169866.1 | - | 3 | 482 | |||
| BC014731.1 | - | 2 | 597 | |||
| BC055447.1 | - | 3 | 536 | 3594406 | ||

Parp16
Genomic Map