siRNA Details
| Gene Symbol: BC003965 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183150.2 | NP_898973.2 | 2 | 200 | ||
| GenBank mRNA | AK141327.1 | - | 1 | 253 | ||
| BC003965.1 | - | 1 | 269 | |||
| BC094389.1 | - | 1 | 271 | |||
| BY262222.1 | - | 2 | 200 | |||
| CF729276.1 | - | 2 | 165 | |||

BC003965
Genomic Map