siRNA Details
| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145991.1 | NP_666103.1 | 16 | 1505 | ||
| GenBank mRNA | AK048027.1 | - | 625 | |||
| AK080861.1 | - | 16 | 1637 | |||
| BC027756.1 | - | 16 | 1606 | 5067496 | ||
| BC031127.1 | - | 16 | 1505 | 4972826 | ||

Cdc73
Genomic Map