siRNA Details
| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145991.1 | NP_666103.1 | 10 | 980 | ||
| GenBank mRNA | AK080861.1 | - | 10 | 1112 | ||
| BC027756.1 | - | 10 | 1081 | 5067496 | ||
| BC031127.1 | - | 10 | 980 | 4972826 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc73| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145991.1 | NP_666103.1 | 10 | 980 | ||
| GenBank mRNA | AK080861.1 | - | 10 | 1112 | ||
| BC027756.1 | - | 10 | 1081 | 5067496 | ||
| BC031127.1 | - | 10 | 980 | 4972826 | ||