siRNA Details
| Gene Symbol: Tmprss13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013373.2 | NP_001013391.2 | 11 | 1439 | ||
| GenBank mRNA | BC010843.1 | - | 3 | 288 | ||
| BC042878.1 | - | 10 | 1175 | |||
| BC085323.1 | - | 11 | 1424 | 30734093 | ||

Tmprss13
Genomic Map