siRNA Details
| Gene Symbol: Cep164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081373.2 | NP_001074842.2 | 8 | 1118 | ||
| GenBank mRNA | AK032890.1 | - | 3 | 279 | ||
| AK220365.1 | - | 4 | 464 | |||
| BC027092.1 | - | 1 | 336 | |||
| BC076567.1 | - | 6 | 859 | |||

Cep164
Genomic Map