siRNA Details
| Gene Symbol: Sidt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289668.1 | NP_001276597.1 | 6 | 1009 | ||
| NM_172257.4 | NP_758461.1 | 6 | 1009 | |||
| GenBank mRNA | AK152371.1 | - | 6 | 942 | ||
| AK155875.1 | - | 6 | 913 | |||
| AK180621.1 | - | 4 | 268 | |||
| BC023957.1 | - | 6 | 1013 | 5321190 | ||
| BC051101.1 | - | 6 | 737 | 5052849 | ||
| CK639655.1 | - | 4 | 340 | |||

Sidt2
Genomic Map