siRNA Details
| Gene Symbol: 4930486L24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178098.2 | NP_835199.1 | 3 | 297 | ||
| GenBank mRNA | AK015635.1 | - | 3 | 267 | ||
| AK145933.1 | - | 3 | 298 | |||
| AY146988.1 | - | 3 | 297 | |||
| BC061218.1 | - | 3 | 284 | 30281034 | ||

4930486L24Rik
Genomic Map