siRNA Details
| Gene Symbol: L3mbtl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289711.1 | NP_001276640.1 | 9 | 1103 | ||
| NM_001289712.1 | NP_001276641.1 | 9 | 1103 | |||
| NM_145993.5 | NP_666105.2 | 9 | 1103 | |||
| GenBank mRNA | AK029115.1 | - | 9 | 1048 | ||
| AK036510.1 | - | 9 | 1056 | |||
| AK132012.1 | - | 9 | 1056 | |||
| AK141810.1 | - | 8 | 2237 | |||
| AK145560.1 | - | 9 | 1045 | |||
| AK146632.1 | - | 9 | 1059 | |||
| AK156236.1 | - | 9 | 1058 | |||
| AY237000.1 | - | 9 | 1036 | |||
| AY237001.1 | - | 9 | 1036 | |||
| AY237002.1 | - | 9 | 1036 | |||
| BC023933.1 | - | 9 | 1044 | 5325131 | ||
| BC030864.1 | - | 9 | 1006 | 4211489 | ||

L3mbtl2
Genomic Map