siRNA Details
Gene Symbol: Chadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164320.1 | NP_001157792.1 | 2 | 177 | ||
GenBank mRNA | AY100452.1 | - | 2 | 233 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164320.1 | NP_001157792.1 | 2 | 177 | ||
GenBank mRNA | AY100452.1 | - | 2 | 233 |