siRNA Details
| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002617.3 | NP_002608.1 | 2, 3 | 261 | ||
| NM_153818.1 | NP_722540.1 | 2, 3 | 261 | |||
| XM_011541573.1 | XP_011539875.1 | 2, 3 | 312 | |||
| XM_011541576.1 | XP_011539878.1 | 2, 3 | 312 | |||
| GenBank mRNA | AB013818.1 | - | 2, 3 | 261 | ||
| AF060502.1 | - | 2, 3 | 224 | |||
| AK124816.1 | - | 2, 3 | 239 | |||
| BC000543.2 | - | 2, 3 | 220 | 3163782 | ||
| BC018198.1 | - | 2, 3 | 243 | 3882695 | ||
| CR542077.1 | - | 2, 3 | 192 | |||
| DA439481.1 | - | 2, 3 | 262 | |||

PEX10
Genomic Map