siRNA Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178115.4 | NP_835216.3 | 24 | 3600 | ||
| GenBank mRNA | AK144179.1 | - | 24 | 3559 | ||
| BC024916.1 | - | 12 | 1732 | |||
| BC046235.1 | - | 12 | 1950 | 6817045 | ||
| BC072596.1 | - | 24 | 3569 | 6409214 | ||

Edrf1
Genomic Map