siRNA Details
| Gene Symbol: Edrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178115.4 | NP_835216.3 | 19 | 2824 | ||
| GenBank mRNA | AK144179.1 | - | 19 | 2783 | ||
| BC024916.1 | - | 7 | 956 | |||
| BC046235.1 | - | 7 | 1174 | 6817045 | ||
| BC072596.1 | - | 19 | 2793 | 6409214 | ||

Edrf1
Genomic Map