siRNA Details
| Gene Symbol: Sertad4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177794.1 | NP_001171265.1 | 4 | 1091 | ||
| NM_198247.2 | NP_937890.2 | 4 | 1346 | |||
| GenBank mRNA | AK047877.1 | - | 1 | 91 | ||
| BC060251.1 | - | 4 | 1315 | 5708574 | ||

Sertad4
Genomic Map