siRNA Details
| Gene Symbol: PEX10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002617.3 | NP_002608.1 | 6 | 1361 | ||
| NM_153818.1 | NP_722540.1 | 6 | 1421 | |||
| XM_011541573.1 | XP_011539875.1 | 6 | 1469 | |||
| GenBank mRNA | AB013818.1 | - | 6 | 1361 | ||
| AK124816.1 | - | 6 | 1339 | |||
| BC000543.2 | - | 6 | 1380 | 3163782 | ||
| BC018198.1 | - | 6 | 1343 | 3882695 | ||

PEX10
Genomic Map