siRNA Details
| Gene Symbol: Syde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166064.1 | NP_001159536.1 | 3 | 2162 | ||
| GenBank mRNA | AK049496.1 | - | 3 | 1380 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Syde2| Gene Symbol: Syde2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166064.1 | NP_001159536.1 | 3 | 2162 | ||
| GenBank mRNA | AK049496.1 | - | 3 | 1380 | ||