siRNA Details
| Gene Symbol: Zfp609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172536.3 | NP_766124.2 | 6 | 3723 | ||
| GenBank mRNA | AK036715.1 | - | 5 | 3456 | ||
| AK129111.1 | - | 2 | 1671 | |||
| BC150683.1 | - | 5 | 3432 | 9087594 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp609| Gene Symbol: Zfp609 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172536.3 | NP_766124.2 | 6 | 3723 | ||
| GenBank mRNA | AK036715.1 | - | 5 | 3456 | ||
| AK129111.1 | - | 2 | 1671 | |||
| BC150683.1 | - | 5 | 3432 | 9087594 | ||