siRNA Details
| Gene Symbol: Neurl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153408.2 | NP_700457.1 | 4 | 919 | ||
| GenBank mRNA | AF321278.1 | - | 4 | 919 | ||
| AK156207.1 | - | 4 | 930 | |||
| BC056622.1 | - | 4 | 910 | 4989283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neurl3| Gene Symbol: Neurl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153408.2 | NP_700457.1 | 4 | 919 | ||
| GenBank mRNA | AF321278.1 | - | 4 | 919 | ||
| AK156207.1 | - | 4 | 930 | |||
| BC056622.1 | - | 4 | 910 | 4989283 | ||