siRNA Details
| Gene Symbol: Lman2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013374.1 | NP_001013392.1 | 4 | 475 | ||
| GenBank mRNA | AK041656.1 | - | 2 | 2320 | ||
| AK142024.1 | - | 444 | ||||
| AK158015.1 | - | 4 | 494 | |||
| BC046969.1 | - | 4 | 475 | 6313679 | ||

Lman2l
Genomic Map